missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-4-nitro-o-cresol, 95%

Product Code. 10260593 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10260593 5 g Glass bottle 5g
10361553 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10260593 Supplier Thermo Scientific Acros Supplier No. 388520050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 772-33-8 | C7H6BrNO3 | 232.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 772-33-8
MDL Number MFCD00007340
InChI Key KFDPCYZHENQOBV-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitrobenzyl bromide, 2-bromomethyl-4-nitrophenol, alpha-bromo-4-nitro-o-cresol, phenol, 2-bromomethyl-4-nitro, koshland reagent i, 2-bromomethyl-4-nitro-phenol, 2-hydroxy-5-nitrobenzylbromide, .alpha.-bromo-4-nitro-o-cresol, o-cresol, .alpha.-bromo-4-nitro, koshland i
PubChem CID 13044
IUPAC Name 2-(bromomethyl)-4-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])CBr)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 143°C to 149°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C7H6BrNO3
Quantity 5 g
Molecular Weight (g/mol) 232.033
Formula Weight 232.03
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material α-Bromo-4-nitro-o-cresol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a-Bromo-4-nitro-o-cresol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.