missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromocinnamaldehyde, 98%

Product Code. 11472337
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472337 25 g 25g
11482337 100 g 100g
11492337 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472337 Supplier Thermo Scientific Alfa Aesar Supplier No. A14230.14
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5443-49-2 | C9H7BrO | 211.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5443-49-2
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD00006965
InChI Key WQRWNOKNRHCLHV-TWGQIWQCSA-N
Synonym alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo
PubChem CID 5369403
IUPAC Name (Z)-2-bromo-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=C(C=O)Br

Specifications

Melting Point 68°C to 72°C
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 211.06
Percent Purity 98%
Chemical Name or Material alpha-Bromocinnamaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.