Learn More
Invitrogen™ alpha-Cardiac Actin Monoclonal Antibody (959606)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA562017
Product Code. 30534834
  • 509.00 € / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
ACTC1 are part of actins, which are highly conserved proteins that are involved in various types of cell motility. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of a two-stranded helix. Each actin can bind to four others. The protein encoded by this gene belongs to the actin family which is comprised of three main groups of actin isoforms, alpha, beta, and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. Defects in this gene have been associated with idiopathic dilated cardiomyopathy (IDC) and familial hypertrophic cardiomyopathy (FHC).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Cardiac Actin
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P68032, P68033, P68035
ACTC1
E.coli-derived recombinant human alpha -Cardiac Actin, Asp3-Phe377.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
959606
Unconjugated
ACTC1
ACTC; Actc1; Actc-1; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; alpha-actin cardiac; alphac-actin; alpha-cardiac actin; ASD5; B-actin; cardiac muscle alpha actin 1; CMD1R; CMH11; LVNC4
Mouse
Protein A/G
RUO
11464, 29275, 70
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized