missing translation for 'onlineSavingsMsg'
Learn More

Alpha-(Chloromethyl)-3-hydroxybenzenemethanol, TRC

Product Code. 30135244
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135244 1 g 1g
30007577 500 mg 500mg
30012913 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135244 Supplier Toronto Research Chemicals Supplier No. TRCC4729501G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1378757-22-2
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.61
Synonym α-(Chloromethyl)-3-hydroxybenzenemethanol, (RS)-2-Chloro-1-(3-hydroxyphenyl)ethanol
IUPAC Name 3-(2-chloro-1-hydroxyethyl)phenol
SMILES OC(CCl)c1cccc(O)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H9ClO2/c9-5-8(11)6-2-1-3-7(10)4-6/h1-4,8,10-11H,5H2
Formula Weight 172.0291
Chemical Name or Material alpha-(Chloromethyl)-3-hydroxybenzenemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.