missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Cortolone, TRC

Product Code. 30011700
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011700 2.5 mg 2.5mg
30138017 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011700 Supplier Toronto Research Chemicals Supplier No. TRCC6962802.5MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-42-7
Molecular Formula C21 H34 O5
Molecular Weight (g/mol) 366.49
Synonym 3α,17,20α,21-Tetrahydroxy-5β-pregnan-11-one, 20α-Cortolone, 3α,17,20α,21-Tetrahydroxy-5β-pregnan-11-one, 3α,17α,20α,21-Tetrahydroxy-5β-pregnan-11-one, 5β-Pregnan-3α,17α,20α,21-tetrol-11-one, 5β-Pregnane-3α,17,20α,21-tetrol-11-one, 5β-Pregnane-3α,17α,20α,21-tetrol-11-one, Cortolon, Cortolone, NSC 59872
IUPAC Name (3R,5R,8S,9S,10S,13S,14S,17R)-17-[(1S)-1,2-dihydroxyethyl]-3,17-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-11-one
SMILES C[C@]12CC[C@@H](O)C[C@H]1CC[C@H]3[C@@H]4CC[C@](O)([C@@H](O)CO)[C@@]4(C)CC(=O)[C@H]23

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C21H34O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12-15,17-18,22-23,25-26H,3-11H2,1-2H3/t12-,13-,14+,15+,17+,18-,19+,20+,21+/m1/s1
Formula Weight 366.2406
Chemical Name or Material alpha-Cortolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.