missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Cyano-4-hydroxycinnamic Acid, TRC

Product Code. 30355176
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30355176 2.5 g 2.5g
30352556 5 g 5g
30354547 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30355176 Supplier Toronto Research Chemicals Supplier No. TRCC3813482.5G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28166-41-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
Synonym 2-Cyano-3-(4-hydroxyphenyl)-2-propenoic Acid, 4-Hydroxy-alpha-cyanocinnamic Acid, NSC 173138, alpha-Cyano-4-hydroxycinnamic Acid
IUPAC Name (E)-2-cyano-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)C(=Cc1ccc(O)cc1)C#N

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C10H7NO3/c11-6-8(10(13)14)5-7-1-3-9(12)4-2-7/h1-5,12H,(H,13,14)/b8-5+
Formula Weight 189.0426
Chemical Name or Material alpha-Cyano-4-hydroxycinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.