missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin hydrate, Thermo Scientific™

Product Code. 11479236
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479236 5 g 5g
11489236 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479236 Supplier Thermo Scientific Alfa Aesar Supplier No. A18092.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
alpha-Cyclodextrin hydrate is used in the food industry. The structure of cyclodextrins allows them to form inclusion complexes with a wide variety of molecules. It lead to enzyme-like specificity in reactions.

Solubility
Soluble in water. 145 g/L at 25

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store in the dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51211-51-9
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.85
MDL Number MFCD00150804
InChI Key HFHDHCJBZVLPGP-UHFFFAOYNA-N
Synonym alpha-cyclodextrin hydrate, water.beta-cyclodextrin, cyclohexaamylose hydrate, ^a-cyclodextrin hydrate, alpha-cyclodextrin water, 972.86 anhydrous
PubChem CID 60161466
IUPAC Name 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Melting Point 279°C (decomposition)
Quantity 5 g
Beilstein 4227442
Merck Index 14,2718
Solubility Information Soluble in water. 145g/L at 25
Optical Rotation +136° (c=10 in Water)
Formula Weight 972.86 (Anhydrous)
Chemical Name or Material alpha-Cyclodextrin hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.