missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Endosulfine Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92259-0.02ml
This item is not returnable.
View return policy
Description
alpha Endosulfine Polyclonal antibody specifically detects alpha Endosulfine in Mouse samples. It is validated for Western Blot
Specifications
| alpha Endosulfine | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| alpha-endosulfine, ARPP-19eMGC4319, endosulfine alpha, MGC78563, MGC8394 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ENSA (NP_996929.1). MAGGLGCDVCYWFVEDTQEKEGILPERAEEAKLKAKYPSLGQKPGGSDFLMKRLQKGQKYFDSGDYNMAKAKMKNKQLPSAGPDKNLVTGDHIPTPQDLP | |
| 0.02 mL | |
| Neuroscience | |
| 2029 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction