missing translation for 'onlineSavingsMsg'
Learn More

alpha-Estradiol 3-Benzoate, TRC

Product Code. 30374464
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374464 500 mg
30380469 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374464 Supplier Toronto Research Chemicals Supplier No. TRCE668605500MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6045-53-0
Molecular Formula C25 H28 O3
Molecular Weight (g/mol) 376.49
Synonym 17alpha-Hydroxyestra-1,3,5(10)-trien-3-yl Benzoate, Estradiol Benzoate Imp. E (EP), 17-epi-Estradiol Benzoate, Estra-1,3,5(10)-triene-3,17-diol, 3-benzoate, (17α)- (9CI, ACI), 17α-Estradiol, 3-benzoate (6CI, 8CI), (+)-17α-Estradiol 3-benzoate, 3-Benzoyloxy-17α-hydroxyoestra-1,3,5(10)-triene, α-Estradiol 3-benzoate, (17α)-Estra-1,3,5(10)-triene-3,17-diol 3-benzoate
IUPAC Name [(8R,9S,13S,14S,17R)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] benzoate
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)c5ccccc5)ccc34)[C@@H]1CC[C@H]2O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23-,25+/m1/s1
Formula Weight 376.2038
Chemical Name or Material 17-epi-Estradiol Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.