missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Estradiol, MedChemExpress

Product Code. 16498126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498126 100 mg 100mg
16412046 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498126 Supplier Medchem Express Supplier No. HYB0141A
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.

Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.

Specifications

Chemical Name or Material Alpha-Estradiol
CAS 57-91-0
Color White
Purity Grade Notes Research
Molecular Formula C18H24O2
Quantity 100 mg
Synonym Alfatradiol Epiestradiol Epiestrol
Solubility Information DMSO : 62.5 mg/mL (229.46 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]12[C@H](O)CC[C@@]1([H])[C@]3([H])CCC4=C(C=CC(O)=C4)[C@@]3([H])CC2
Molecular Weight (g/mol) 272.38
Formula Weight 272.38
Percent Purity 99.52%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.