Learn More
alpha-Fetoprotein/AFP Antibody (CL9766) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21173UV
Product Code. 30308595
  • 737.00 € / 0.10mL
Description
alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Fetoprotein/AFP
Monoclonal
DyLight 350
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
This antibody was developed using a recombinant protein derived from P02771, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
174
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9766
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified