missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Hexylcinnamaldehyde, TRC

Product Code. 30396555
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396555 100 g -
30395564 500 g 500g
30392109 250 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396555 Supplier Toronto Research Chemicals Supplier No. TRCH295705100G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-86-0
Molecular Formula C15 H20 O
Molecular Weight (g/mol) 216.32
Synonym Hexylcinnamal, Octanal, 2-(phenylmethylene)- (9CI, ACI), 2-(Phenylmethylene)octanal (ACI), Cinnamaldehyde, α-hexyl- (6CI, 8CI), 2-Benzylideneoctanal, 2-Hexyl-3-phenyl-2-propenal, 2-Hexylcinnamaldehyde, Hexyl cinnamal, Hexyl cinnamic aldehyde, NSC 406799, NSC 46150, α-Hexylcinnamaldehyde, α-Hexylcinnamic aldehyde, α-Hexylcinnamyl aldehyde, α-n-Hexyl-β-phenylacrolein, α-n-Hexylcinnamaldehyde
IUPAC Name (2E)-2-benzylideneoctanal
SMILES CCCCCC\C(=C/c1ccccc1)\C=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C15H20O/c1-2-3-4-6-11-15(13-16)12-14-9-7-5-8-10-14/h5,7-10,12-13H,2-4,6,11H2,1H3/b15-12+
Formula Weight 216.1514
Chemical Name or Material Hexylcinnamaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.