missing translation for 'onlineSavingsMsg'
Learn More

alpha-Isopropylbenzylamine Hydrochloride, TRC

Product Code. 30133151
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133151 500 mg 500mg
30012096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133151 Supplier Toronto Research Chemicals Supplier No. TRCI822815500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24290-47-9
Molecular Formula C10H15N•HCl
Molecular Weight (g/mol) 149.233646
Synonym (2-Methyl-1-phenylpropyl)amine Hydrochloride, α-(1-Methylethyl)Benzenemethanamine Hydrochloride, α-Isopropylbenzylamine Hydrochloride, α-Isopropyl-benzylamine Hydrochloride;
IUPAC Name 2-methyl-1-phenylpropan-1-amine;hydrochloride
SMILES CC(C)C(C1=CC=CC=C1)N.Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H15N.ClH/c1-8(2)10(11)9-6-4-3-5-7-9;/h3-8,10H,11H2,1-2H3;1H
Formula Weight 185.0971
Chemical Name or Material alpha-Isopropylbenzylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.