missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Methoxy Albuterol, TRC

Product Code. 30003974
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003974 25 mg 25mg
30003919 1 mg 1mg
30004524 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003974 Supplier Toronto Research Chemicals Supplier No. TRCM22605525MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870076-72-5
Molecular Formula C14 H23 N O3
Molecular Weight (g/mol) 253.34
Synonym Benzenemethanol, 5-[2-[(1,1-dimethylethyl)amino]-1-methoxyethyl]-2-hydroxy-, 4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol, Salbutamol Sulphate Imp A (EP), [5-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxyphenyl]methanol
IUPAC Name 4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
SMILES COC(CNC(C)(C)C)c1ccc(O)c(CO)c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H23NO3/c1-14(2,3)15-8-13(18-4)10-5-6-12(17)11(7-10)9-16/h5-7,13,15-17H,8-9H2,1-4H3
Formula Weight 253.1678
Chemical Name or Material 4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.