Learn More
alpha-Methylacyl-CoA Racemase/AMACR Antibody (CL9362) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88931AF750
Product Code. 30292435
  • 737.00 € / 0.10mL
Description
alpha-Methylacyl-CoA Racemase/AMACR Monoclonal antibody specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Methylacyl-CoA Racemase/AMACR
Monoclonal
Alexa Fluor 750
2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM
This antibody was developed using a recombinant protein derived from Q9UHK6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Lipid and Metabolism, Prostate Cancer, Signal Transduction
23600
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9362
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified