missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylcinnamic acid, 99%

Product Code. 11350234
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11350234 100 g 100g
11330234 5 g 5g
11340234 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11350234

Brand: Thermo Scientific Alfa Aesar L00806.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1199-77-5 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1199-77-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00002652
InChI Key XNCRUNXWPDJHGV-BQYQJAHWSA-N
Synonym alpha-methylcinnamic acid, 2-methyl-3-phenylacrylic acid, 2e-2-methyl-3-phenylprop-2-enoic acid, a-methylcinnamic acid, 2e-2-methyl-3-phenylacrylic acid, .alpha.-methylcinnamic acid, e-2-methyl-3-phenylacrylic acid, 2-propenoic acid, 2-methyl-3-phenyl, alpha-methyl cinnamic acid, e-2-methyl-3-phenylprop-2-enoic acid
PubChem CID 637817
IUPAC Name (E)-2-methyl-3-phenylprop-2-enoic acid
SMILES C\C(=C/C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 78°C to 82°C
Boiling Point 288°C
Quantity 100 g
Beilstein 2042544
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material alpha-Methylcinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.