missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylstyrene, 99%, stab. with 10-20 ppm 4-tert-butylcatechol

Product Code. 11362236
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362236 2500 mL 2.5 L
11342236 100 mL 100mL
11352236 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362236 Supplier Thermo Scientific Alfa Aesar Supplier No. L03609.0F
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-83-9 | C9H10 | 118.18 g/mol

α-Methylstyrene (AMS) is used in the synthesis of (acrylonitrile-styrene-acrylic copolymer)/(α-methylstyrene-acrylonitrile copolymer). It is used in the manufacture of plasticizers, resins and polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Methylstyrene (AMS) is used in the synthesis of (acrylonitrile-styrene-acrylic copolymer)/(α-methylstyrene-acrylonitrile copolymer). It is used in the manufacture of plasticizers, resins and polymers.

Solubility
Soluble in water.( 0,1 g/L) at 25°C.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents, Peroxides, Organometallic compounds, Metallic salts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-83-9
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008859
InChI Key XYLMUPLGERFSHI-UHFFFAOYSA-N
Synonym alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene
PubChem CID 7407
ChEBI CHEBI:35060
IUPAC Name prop-1-en-2-ylbenzene
SMILES CC(=C)C1=CC=CC=C1

Specifications

Melting Point -23°C
Density 0.909
Boiling Point 166°C to 169°C
Flash Point 45°C (113°F)
Refractive Index 1.538
Quantity 2500 mL
UN Number UN2303
Beilstein 969405
Solubility Information Soluble in water.( 0,1g/L) at 25°C.
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material alpha-Methylstyrene, stabilized with 10 to 20ppm 4-tert-butylcatechol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • .alpha.-Methylstyrene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: .alpha.-Methylstyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.