missing translation for 'onlineSavingsMsg'
Learn More

alpha-Naphtholbenzein

Product Code. 11493157
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493157 25 g 25g
11483157 5 g 5g
11403167 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493157 Supplier Thermo Scientific Alfa Aesar Supplier No. A16125.14
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 145-50-6 | C27H18O2 | 374.439 g/mol

α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

Solubility
Soluble in ethanol, acetic acid and acetone. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, water reactive substances, azo, diazo, hydrazines, metals, nitrides, peroxides, hydroperoxides, epoxides and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-50-6
Molecular Formula C27H18O2
Molecular Weight (g/mol) 374.439
MDL Number MFCD00078492
InChI Key VDDWRTZCUJCDJM-PNHLSOANSA-N
PubChem CID 5941340
IUPAC Name (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one
SMILES C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O

Specifications

Melting Point ∼225°C (decomposition)
Quantity 25 g
Beilstein 3471575
Synonym p-naphtholbenzein, 1-naphtholbenzein, alpha-naphtholbenzein, .alpha.-naphtholbenzein, 4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one, naphtholbenzein, 1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene
Solubility Information Soluble in ethanol,acetic acid and acetone. Insoluble in water.
Formula Weight 374.44
Chemical Name or Material alpha-Naphtholbenzein
compliance-icons
Product Identifier
  • alpha-Naphtholbenzein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: alpha-Naphtholbenzein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.