missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-parvin/Actopaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00222-50ul
This item is not returnable.
View return policy
Description
Alpha-parvin/Actopaxin Polyclonal antibody specifically detects Alpha-parvin/Actopaxin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Alpha-parvin/Actopaxin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| actopaxin, alpha-parvin, Calponin-like integrin-linked kinase-binding protein, CH-ILKBP, FLJ10793, FLJ12254, FLJ25106, FLJ46049, Matrix-remodeling-associated protein 2, matrix-remodelling associated 2, MXRA2, parvin, alpha | |
| Produced in rabbits immunized with purified, recombinant Human Alpha-parvin/Actopaxin (Uniprot#: Q9NVD7-1; Met1-Glu372). | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55742 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction