missing translation for 'onlineSavingsMsg'
Learn More

Skytec™ Alpha Sentinel Premium Reusable Half Face Mask

Product Code. 30242991 Shop All Skytec Products
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30242991 Small Each
30242978 Medium Each
30242993 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30242991 Supplier Skytec™ Supplier No. ASRAS0001BD
  • ONLINE EXCLUSIVE PRICE
    48.77 €
    32.47 € / Pack of 1
    Save 16.30 €
    33% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Alpha Sentinel Half Masks are ergonomically designed for optimum protection, comfort and PPE compatibility.

  • Well-balanced and lightweight (140 g), providing optimum wearer comfort
  • Available in three sizes, creating a tailored fit profile, superb fit and comfort for any workforce
  • Fully adjustable head harness, offering easy fitting and adjustment, with a robust and trusted fit when in use
  • Low profile shape to maximise the field of vision and allow exceptional compatibility with other PPE
  • Alpha Sentinel filters are fitted with a simple connection which easily clicks into position
  • Premium quality elastic strap provides a secure high comfort fit and retains mask positioning
  • The large exhalation valve reduces heat and moisture build-up, providing low breathing resistance
  • Easily cleaned and maintained with spare parts available
  • Quantitative face fit adapter available
  • Respirators are supplied with user seal check covers, which double as particulate retainers for combination filter

Specifications

Certifications/Compliance CE , UKCA , EN140
Color Black
Disposable Reusable
Filter Type HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA
Material Thermoplastic Elastomer
Product Type Half Mask Respirator
Size Small
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Filter dependent: Toxic Waste Disposal, Metal Cutting and Casting, Aerospace, Petrochemical
For Use With (Equipment) Alpha Sentinel Filters
Harness Material Thermoplastic Elastomer
Harness Style Standard
Latex Free Yes
Material Headstrap Polyester
Model Alpha Sentinel Half Mask
Neck Strap Polyester
Packaging Type Box
Quantity 16/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.