missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Mouse, Clone: 1A4/asm-1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 23 publications
245.00 € - 754.00 €
Specifications
| Antigen | alpha-Smooth Muscle Actin |
|---|---|
| Clone | 1A4/asm-1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-1ug/ml, Simple Western 10 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 1:600 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18738943
|
Novus Biologicals
NBP2-33006-0.02mg |
0.02 mg |
245.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758803
|
Novus Biologicals
NBP2-33006-0.1mg |
0.1 mg |
521.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18084824
|
Novus Biologicals
NBP2-33006-0.2MG |
0.2 mg |
754.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Baboon, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
| alpha-Smooth Muscle Actin | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Pig, Baboon, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Monkey, Rabbit, Sheep | |
| P62736 | |
| 59 | |
| N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH | |
| Primary | |
| Store at 4C. | |
| 42 kDa |
| 1A4/asm-1 | |
| Western Blot 0.5-1ug/ml, Simple Western 10 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 1:600 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 1a4 sma, 1a4 smooth muscle actin, AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTVS, alpha 2 actin, alpha SMA, alpha sma 1a4, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, sma 1a4, smooth muscle actin | |
| ACTA2 | |
| IgG2a κ | |
| Protein A purified | |
| Actin is a major component of the cytoskeleton and is present in most cell types. This MAb is highly specific to actin from smooth muscles. Its epitope lies in the first four N-terminal amino acids. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actin and anti-smooth muscle actin and are negative with anti-myogenin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title