missing translation for 'onlineSavingsMsg'
Learn More

alpha Tubulin Mouse anti-Human, Mouse, Rat, HRP, Clone: 1E4C11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech HRP-66031-100UL

Product Code. 16801256

  • 189.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predomitly found in morphologically differentiated neurologic cells.
Specifications

Specifications

alpha Tubulin
Monoclonal
1.33 mg/mL
PBS with 50% glycerol and 0.02% thimerosal; pH 7.3
P05213, P68363, Q6P9V9
Tuba1b
Antigen Affinity Chromatography
RUO
10376, 22143, 500929
-20°C
Liquid
Western Blot
1E4C11
HRP
TUBA1B
Alpha tubulin ubiquitous, alpha tubulin,a tubulin, HRP Tubulin Alpha, K ALPHA 1, TUBA1B, tubulin, Tubulin Alpha, Tubulin alpha 1B chain, Tubulin alpha ubiquitous chain, Tubulin K alpha 1, tubulin, alpha 1b
Mouse
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.