missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB120-13497-0.025ml
This item is not returnable.
View return policy
Description
AlphaB Crystallin/CRYAB Polyclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AlphaB Crystallin/CRYAB | |
| Polyclonal | |
| Western Blot 1:5000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_001876.1 | |
| CRYAB | |
| Synthetic peptide corresponding to human alpha B crystallin conjugated to KLH | |
| 0.025 mL | |
| Neuroscience, Vision | |
| 1410 | |
| Human | |
| Antisera |
| Western Blot | |
| Unconjugated | |
| PBS pH 7.4, 50% Glycerol with 0.09% Sodium Azide | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Detects approx 22kDa. Does not cross-react with alpha A-crystallin. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction