missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHX8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568529
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
Members of the LIM homeobox gene family, such as LHX8, encode transcription regulators that share common structural features. They all contain 2 tandemly repeated cysteine-rich double-zinc finger motifs, called LIM domains, in addition to a homeodomain. The homeodomain is a DNA-binding domain, and the LIM domains are essential for regulating the activity of these molecules by interacting with other proteins. Members of the LIM homeobox gene family are required for the patterning or the specification and differentiation of different cell types during embryonic development (Zhao et al., 1999 [PubMed 10611327]).
Specifications
| LHX8 | |
| Polyclonal | |
| Unconjugated | |
| LHX8 | |
| L3; Lhx7; LHX8; LIM homeo box protein 8; LIM homeobox 8; LIM homeobox protein 7; LIM homeobox protein 8; LIM Homeodomain 8; LIM/homeobox protein Lhx7; LIM/homeobox protein Lhx8; LIM-homeodomain protein Lhx8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 365963 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| LHX8 | |
| synthetic peptide directed towards the N-terminal region of rat LHX8. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction