missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-90185-25ul
This item is not returnable.
View return policy
Description
ALS2CR15 Polyclonal specifically detects ALS2CR15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ALS2CR15 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q8NDH6 | |
| ICA1L | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SPSASLTSQEPSMGSEPLAHSSRFLPSQLFDLGFHVAGAFNNWVSQEESELCLSHTDNQPVPSQSPKKLTRSPNNGNQD | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ALS2CR14, ALS2CR15, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 14, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 14 protein, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein, candidate 15, DKFZp434E1919, Ica69-related protein, islet cell autoantigen 1,69kDa-like, islet cell autoantigen 1-like protein, MGC138440 | |
| Rabbit | |
| 54 kDa | |
| 25 μL | |
| Signal Transduction | |
| 130026 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction