missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR8 Antibody (S04-5G7), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 € - 645.00 €
Specifications
| Antigen | ALS2CR8 |
|---|---|
| Clone | S04-5G7 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680145
|
Novus Biologicals
NBP3-19568-100ul |
100 μg |
645.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601194
|
Novus Biologicals
NBP3-19568-50ul |
50 μg |
421.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALS2CR8 Monoclonal antibody specifically detects ALS2CR8 in Human samples. It is validated for Western BlotSpecifications
| ALS2CR8 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 79800 | |
| IgG | |
| Affinity purified |
| S04-5G7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 8, amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein, Calcium-response factor, CaRF, CARFDKFZp667N246, FLJ12675, FLJ21579, NYD-SP24, Testis development protein NYD-SP24 | |
| Recombinant protein of human ALS2CR8 (Uniprot # Q8N187) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title