missing translation for 'onlineSavingsMsg'
Learn More

Aluminon, ACS

Product Code. 11343298
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343298 100 g 100g
11333298 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343298 Supplier Thermo Scientific Alfa Aesar Supplier No. 036239.22
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 569-58-4 | C22H23N3O9 | 473.438 g/mol

It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-58-4
Molecular Formula C22H23N3O9
Molecular Weight (g/mol) 473.438
MDL Number MFCD00040925
InChI Key AIPNSHNRCQOTRI-UHFFFAOYSA-N
Synonym aluminon, ammonium aurintricarboxylate, aurintricarboxylic acid ammonium salt, aurintricarboxylic acid, acs
PubChem CID 54729869
ChEBI CHEBI:87398
IUPAC Name triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]

Specifications

Melting Point 220°C to 225°C (decomposition)
pH 5.3
Odor Odorless
Quantity 100 g
Merck Index 14,322
Solubility Information Soluble in water(800g/L).
Formula Weight 473.44
Grade ACS
Physical Form Powder
Chemical Name or Material Aluminon
compliance-icons
Product Identifier
  • Aluminon, ACS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Aluminon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.