missing translation for 'onlineSavingsMsg'
Learn More

Aluminum hydroxide, extra pure, powder

Product Code. 10328070 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
missing translation for 'unitSize'
10kg
25g
2kg
500g
This item is not returnable. View return policy

Product Code. 10328070

missing translation for 'mfr': Thermo Scientific Acros 219130020

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]

Specifications

Color White
pH 8.5 to 10 (5% aq. suspension)
Physical Form Powder
Quantity 2 kg
Linear Formula Al(OH)3
Merck Index 15,338
Solubility Information Solubility in water: insoluble
Formula Weight 78
Percent Purity ≥63.5% (Al2O3)
Grade Extra Pure
Loss on Ignition 36.5% max.
Packaging Plastic bottle
Chemical Name or Material Aluminum hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium hydroxide
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.