missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Hydroxide, Technical Grade, TRC

Product Code. 30006112
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006112 25 g 25g
30007093 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006112 Supplier Toronto Research Chemicals Supplier No. TRCA57574525G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21645-51-2
Molecular Formula Al H3 O3
Molecular Weight (g/mol) 78
Synonym Aluminium Hydroxide, Aluminum hydroxide (Al(OH)3), Aluminum hydroxide (6CI,8CI), Alumina hydrate, Alumina trihydrate, Aluminic acid (H3AlO3), Aluminum hydroxide gel, Aluminum oxide (Al2O3), trihydrate, Aluminum oxide hydrate, Aluminum oxide trihydrate, Aluminum trihydroxide, Alusal, Aluxid, Amberol ST 140F, Amphogel, Amphojel, Antipollon HT, Apyral, Hydralcoat, Hydrated alumina, Hydrated aluminum oxide, Hydroxal, Trihydroxyaluminum
IUPAC Name trihydroxyalumane
SMILES O[Al](O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/Al.3H2O/h;3*1H2/q+3;;;/p-3
Formula Weight 77.9898
Chemical Name or Material Aluminium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.