Learn More
Merck Aluminum MultiScreen Column Loader
Product Code. 10106680 Shop All Merck Products
Change view
Click to view available options
Total Volume (Metric) Well:
80 μL
Unit Size:
Each
Product Code. Total Volume (Metric) Well Unit Size
10106680 80 μL Each
1 options
Product Code. 10106680 Supplier Merck Supplier No. MACL09680
  • ONLINE EXCLUSIVE PRICE
    791.00 €
    728.08 € / Pack of 1
    Save 62.92 €
    7% Off
Estimated Shipment: 12-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High throughput MultiScreen™HTS filter plates are specifically developed for use with automation
Fully Compatible with Automation
High throughput MultiScreen™HTS filter plates are specifically developed for use with automation. The 96 well and 384 well plates are designed to standardised dimensions and meet ANSI/SBS 2004 plate specifications. They are fully compatible with automated gripper arms, stackers, barcode labels, and plate readers.
Innovative Plate Design
Rigid sidewalls are aligned for easy handling by robotic gripper arm and also provide ample surfaces for bar code labels. The plate design protects the underdrain and eliminates surface contact with individual well 'drip directors'. The plastic skirt enables stacking directly with collection plates and improves the vacuum seal when used with a Millipore™ vacuum manifold. The removable underdrain gives full access to membranes and is ideal for applications including Elispot, radioisotope detection and whole organism visualisation.
Extensive Product Range
MultiScreen™HTS filter plates are available in a broad range of Millipore plastics and membranes. MultiScreen™ Solubility filter plates are also optimised in HTS format for filtration-based solubility determination. Accessory column loaders are available for loading chromatography media for column separators. MultiScreen™HTS filter plates are available in clear, opaque and white. For non-standard plastic and membrane combinations, custom requests are accepted. Available membranes include:
  • Hydrophilic Durapore™ PVDF membrane
  • Glass fibre
  • Hydrophobic Immobilon™-P membrane
  • HA mixed cellulose esters
  • Negatively charged phosphocellulose
  • Positively charged DEAE
  • Track-etched polycarbonate Applications include: receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation and radioisotope detection.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Black
Material Aluminum
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 80 μL
Quantity 1/Pk.