missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL

Product Code. 11351429
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351429 50 mL 50mL
11341429 100 mL 100mL
11480360 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11351429 Supplier Thermo Scientific Alfa Aesar Supplier No. 013856.AD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7429-90-5 , 7732-18-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifica

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Type Analytical Standard
IUPAC Name aluminium
Quantity 50 mL
Grade Specpure
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 5.0%; Aluminum: 0.1%; Water: 94.9%
Chemical Name or Material Aluminum
compliance-icons
Identificativo prodotto
  • Aluminum, plasma standard solution, Specpure Al 1000µg/ml
Avvertenza
  • Danger
Categoria di pericolo
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
Consigli di prudenza
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Informazioni supplementari
  • MIXTURE LIST-Contain: Hydrochloric acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.