Learn More
Aluminum sec-butoxide, 95%
Description
It is used for active catalyst and support, inorganic membrane, redrcing agent of aldehyde-ketone, oxidant of aldehyde-ketone, gel forming agent of printing ink and paint, dehydrant, water proofing agent and aluminum base grease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.967 g/mL |
| Boiling Point | 200°C to 206°C (30 mmHg) |
| Flash Point | 27°C (82°F) |
| Odor | Alcohol-like |
| Refractive Index | 1.439 |
| Quantity | 10 g |
| UN Number | UN1992 |
| Beilstein | 3910908 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Hydrolyzes in water; Soluble in aromatic hydrocarbons, alcohols |
| Show More |
Safety and Handling
- Aluminum sec-butoxide
- Warning
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.