missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate Hydrate, 98%, Honeywell™ Fluka™

Product Code. p-7120155 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
Product Code. Quantity Packaging unitSize
15615150 500 g Poly Bottle 500g
15605150 2.5 kg Poly Bottle 2.5kg
2 options
This item is not returnable. View return policy

Product Code. 15615150

Brand: Honeywell-Fluka 368458500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

>98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17927-65-0
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.13
MDL Number MFCD00149136
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o
PubChem CID 3080712
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 86.5°C
Color White
Quantity 500 g
Linear Formula Al2(SO4)3 · xH2O
UN Number UN3260
IUPAC Name dialuminium(3+) trisulfate
Formula Weight 342.15g/mol (anhydrous basis)
Percent Purity 98%
Packaging Poly Bottle
Appearance Solid
Density 1.69g/mL at 25°C
Chemical Name or Material Aluminum sulfate hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.