missing translation for 'onlineSavingsMsg'
Learn More

Alverine citrate, MedChemExpress

Product Code. 16460958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460958 5 g 5g
16458508 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460958 Supplier Medchem Express Supplier No. HYB0500
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Alverine citrate is a 5-HT1A receptor antagonist, with an IC50 of 101 nM.

Alverine citrate is a 5-HT1A receptor antagonist, with an IC50 of 101 nM.

Specifications

Chemical Name or Material Alverine citrate
Physical Form Solid
CAS 5560-59-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C26H35NO7
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym NSC 35459
Solubility Information DMSO : 100 mg/mL (211.17 mM; Need ultrasonic)
SMILES CCN(CCCC1=CC=CC=C1)CCCC2=CC=CC=C2.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 473.56
Formula Weight 473.56
Percent Purity 99.43%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.