missing translation for 'onlineSavingsMsg'
Learn More

Alverine, MP Biomedicals™

Product Code. 11444771 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444771 25 g 25g
11434771 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444771 Supplier MP Biomedicals Supplier No. 0215473225
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5560-59-8
Molecular Formula C26H35NO7
Molecular Weight (g/mol) 473.566
InChI Key RYHCACJBKCOBTJ-UHFFFAOYSA-N
Synonym alverine citrate, alverine citrate salt, nci 85x, unii-9jfb58yk1e, 9jfb58yk1e, n-ethyl-3,3'-diphenyldipropylamine citrate 1:1, dsstox_cid_25562, dsstox_rid_80959, dsstox_gsid_45562
PubChem CID 21718
ChEBI CHEBI:53785
IUPAC Name N-ethyl-3-phenyl-N-(3-phenylpropyl)propan-1-amine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CCCC1=CC=CC=C1)CCCC2=CC=CC=C2.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Quantity 25 g
Formula Weight 473.6
Physical Form Powder
Chemical Name or Material Alverine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.