missing translation for 'onlineSavingsMsg'
Learn More

AM 580

Product Code. 15476759
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476759 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15476759 Supplier Thermo Scientific Alfa Aesar Supplier No. J65155.MA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102121-60-8 | C22H25NO3 | 351.45 g/mol

Retinoid derivative used for gene expression studies of leukemia-retinoic acid receptor (PHL-RAR) cell line differentiation. AM 580 inhibits tumor proliferation by reducing the level of RARγ and activates RAR β. This receptor increases the expression of disintegrin and metalloprotease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Retinoid derivative used for gene expression studies of leukemia-retinoic acid receptor (PHL-RAR) cell line differentiation. AM 580 inhibits tumor proliferation by reducing the level of RARγ and activates RAR β. This recep
TRUSTED_SUSTAINABILITY

Specifications

CAS 102121-60-8
Color White
Molecular Formula C22H25NO3
MDL Number MFCD00673916
Quantity 10 mg
Synonym unii-vz5s5g9zfz, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl carbonyl amino benzoic acid, vz5s5g9zfz, chembl69367, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxamido benzoic acid, benzoic acid,4-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl carbonyl amino, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl carbonyl aminobenzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl carboxamido benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl carbonyl amino benzoic acid, benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl carbonyl amino
Solubility Information Soluble in DMSO (20mg/ml) or ethanol (10mg/ml).
InChI Key SZWKGOZKRMMLAJ-UHFFFAOYSA-N
SMILES CC1(C)CCC(C)(C)C2=CC(=CC=C12)C(=O)NC1=CC=C(C=C1)C(O)=O
IUPAC Name 4-[(5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-carbonyl)amino]benzoic acid
Molecular Weight (g/mol) 351.45
PubChem CID 2126
ChEBI CHEBI:64210
Formula Weight 351.44
Physical Form Crystalline Solid
Chemical Name or Material AM 580
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.