missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AM 80

Product Code. 18095392
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095392 10 mg 10mg
18026002 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095392 Supplier R&D Systems™ Supplier No. 3507/10
  • ONLINE EXCLUSIVE PRICE
    233.27 €
    208.95 € / 10mg
    Save 24.32 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. RARα agonist; anticancer agent

Retinoic acid receptor α (RARα) agonist that induces differentiation (ED50 = 0.79 nM) and apoptosis of HL-60 cells in vitro. Exhibits antiproliferative effects against a variety of human tumor cells lines (mean values of 35, 40 and 60% growth inhibition at 0.1, 1 and 10 μM respectively) and displays anticancer activity against acute promyelocytic leukemia in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94497-51-5
Molecular Formula C22H25NO3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 351.44
Purity >99%
Molecular Weight (g/mol) 351.45
Chemical Name or Material AM 80
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.