missing translation for 'onlineSavingsMsg'
Learn More

AM095 free acid, MedChemExpress

Product Code. 16473027
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473027 50 mg 50mg
16467046 100 mg 100mg
16459517 5 mg 5mg
16441216 200 mg 200mg
16411806 500 mg 500mg
16406978 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16473027 Supplier Medchem Express Supplier No. HY16040
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.

AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.

Specifications

Chemical Name or Material AM095 free acid
CAS 1228690-36-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H24N2O5
Quantity 50 mg
Solubility Information DMSO : 67.3 mg/mL (147.43 mM; Need ultrasonic and warming)
SMILES O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1
Molecular Weight (g/mol) 456.49
Formula Weight 456.49
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.