missing translation for 'onlineSavingsMsg'
Learn More

AM211, MedChemExpress

Product Code. 16498257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498257 5 mg 5mg
16445558 10 mg 10mg
16436116 100 mg 100mg
16421727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498257 Supplier Medchem Express Supplier No. HY13213
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.

AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.

Specifications

Chemical Name or Material AM211
CAS 1175526-27-8
Color White
Purity Grade Notes Research
Molecular Formula C27H27F3N2O4
Quantity 5 mg
Synonym AM211 free acid
Solubility Information DMSO : ≥ 125 mg/mL (249.75 mM)
SMILES O=C(O)CC1=CC(C2=CC=C(C(F)(F)F)C=C2CN(CC)C(NCC3=CC=CC=C3)=O)=C(OC)C=C1
Molecular Weight (g/mol) 500.51
Formula Weight 500.51
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.