missing translation for 'onlineSavingsMsg'
Learn More

Amantadine hydrochloride, MedChemExpress

Product Code. 16468478
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468478 10 g 10g
16444357 50 g 50g
16400918 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468478 Supplier Medchem Express Supplier No. HYB0402A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amantadine (1-Adamantanamine) hydrochloride is an orally avtive and potent antiviral agent with activity against influenza A viruses.

Amantadine hydrochloride inhibits several ion channels such as NMDA and M2, and also inhibits Coronavirus ion channels. Amantadine hydrochloride also has anti-orthopoxvirus and anticancer activity. Amantadine hydrochloride can be used for Parkinson's disease, postoperative cognitive dysfunction (POCD) and COVID-19 research.

Specifications

Chemical Name or Material Amantadine hydrochloride
Physical Form Solid
CAS 665-66-7
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C10H18ClN
For Use With (Application) COVID-19-anti-virus
Synonym 1-Adamantanamine hydrochloride 1-Adamantylamine hydrochloride 1-Aminoadamantane hydrochloride
Solubility Information DMSO : 100 mg/mL (532.74 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (266.37 mM)
SMILES NC1(C2)C[C@@H]3C[C@@H](C[C@H]2C3)C1.Cl
Molecular Weight (g/mol) 187.71
Formula Weight 187.71
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.