missing translation for 'onlineSavingsMsg'
Learn More

Amantadine Hydrochloride, TRC

Product Code. 30137158
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137158 5 g 5g
17912465 100 mg 100mg
17914345 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137158 Supplier Toronto Research Chemicals Supplier No. TRCA5758205G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 665-66-7
Molecular Formula C10 H17 N . Cl H
Molecular Weight (g/mol) 187.71
Synonym 1-Adamantanamine, hydrochloride (8CI), Adamantanamine hydrochloride (6CI), Tricyclo[3.3.1.13,7]decan-1-amine, hydrochloride (9CI), 1-Adamantamine hydrochloride, 1-Adamantylamine hydrochloride, 1-Aminoadamantane hydrochloride, ADS 5102, Adamantylamine hydrochloride, Adamin, Adamine, Amantadine hydrochloride, Amazolon, Aminoadamantane hydrochloride, EXP 105-1, Mantadan, Mantadine, Mantadix, Midantan, Midantane, Mydantane, NSC 83653, Symadine, Symmetrel, Virasol, Virofral
IUPAC Name adamantan-1-amine;hydrochloride
SMILES Cl.NC12CC3CC(CC(C3)C1)C2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H17N.ClH/c11-10-4-7-1-8(5-10)3-9(2-7)6-10;/h7-9H,1-6,11H2;1H
Formula Weight 187.1128
Percent Purity >95%
Chemical Name or Material Amantadine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.