missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 10X TBE (powder)

Product Code. 10534835
Click to view available options
Quantity:
10 x 1 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10534835

Brand: Invitrogen™ AM9864

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Invitrogen 10X TBE (Tris-Borate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitive application.

Invitrogen 10X TBE (Tris-Borate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitive application. 10 packets are provided, each containing the necessary powder to make 1 L of 10X TBE buffer upon the addition of H2O. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Running Buffer
Concentration 10X
For Use With (Application) Nucleic acid gel electrophoresis & blotting
Physical Form Powder
Product Line Ambion
Quantity 10 x 1 L

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.