missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ In Vivo Pre-Designed siRNA

Product Code. 15551114
Change view
Click to view available options
Final Product Type:
Ambion™ In Vivo Custom SIRNA HPLC
Ambion™ In Vivo Pre-Designed HPLC IVR
Ambion™ In Vivo Pre-Designed SIRNA HPLC
Ambion™ In Vivo Pre-Designed siRNA
Quantity:
1 μmol
250 nmol
Unit Size:
250µmol
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Final Product Type Quantity Unit Size
15551114 Ambion™ In Vivo Pre-Designed HPLC IVR 1 μmol Each
15501114 Ambion™ In Vivo Custom SIRNA HPLC 250 nmol Each
15286304 Ambion™ In Vivo Pre-Designed siRNA 250 nmol Each
15511114 Ambion™ In Vivo Custom SIRNA HPLC 1 μmol Each
15183197 Ambion™ In Vivo Pre-Designed SIRNA HPLC 250 nmol 250µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15551114 Supplier Invitrogen™ Supplier No. 4457310
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ambion In Vivo Predesigned siRNAs are specifically designed for gene silencing applications in animal models.

Ambion In Vivo Predesigned siRNAs are specifically designed for gene silencing applications in animal models. These siRNAs are chemically modified to enhance stability, reduce immunogenicity, and improve delivery efficiency, making them excellent for in vivo studies. Ambion In Vivo Predesigned siRNAs allow researchers to investigate gene function in a physiological context, offering valuable insights into gene regulation mechanisms and disease processes. Built using the Silencer Select algorithm, these predesigned siRNAs ensure high specificity and efficiency.

Key features include:

  • Enhanced stability—chemical modifications help ensure that Ambion In Vivo Predesigned siRNAs remain stable in the bloodstream and within tissues, helping provide sustained gene silencing effects
  • Reduced immunogenicity—modifications minimize immune responses, helping ensure safe and effective use in animal models
  • Efficient delivery—optimized for use with various delivery systems, including lipid nanoparticles, helping ensure efficient uptake by target cells
  • High specificity—designed to precisely target specific mRNAs, resulting in potent and specific gene knockdown

In vivo-ready siRNA

Ambion In Vivo Predesigned siRNAs are processed to be ready for in vivo use. This includes HPLC purification, membrane dialysis to remove salts and solvents, and sterile filtration prior to drying down.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Description Ambion In Vivo Pre-Designed HPLC IVR, 1 umol
RNAi Type siRNA
Final Product Type Ambion™ In Vivo Pre-Designed HPLC IVR
Product Line Ambion
Product Type siRNA
Purity HPLC
Quantity 1 μmol
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.