missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AMBMP Hydrochloride, >98%
GREENER_CHOICE

Product Code. 30000913
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000913

Brand: Tocris Bioscience™ 6043/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wnt canonical signaling activator; also tubulin polymerization inhibitor

Wnt canonical signaling activator. Also inhibits tubulin polymerization. Suppresses TLR2/4/5-induced inflammatory response in human monocytes. Inhibits cell proliferation and induces cell cycle arrest in MDA-MB-231 breast cancer cells. Also decreases embryo to blastocyst development.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2095432-75-8
Molecular Formula C19H19ClN4O3

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 386.84
Purity >98%
Molecular Weight (g/mol) 386.836
Chemical Name or Material N4-(1,3-Benzodioxol-5-ylmethyl)-6-(3-methoxyphenyl)-2,4-pyrimidinediamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.