missing translation for 'onlineSavingsMsg'
Learn More

Ambrisentan, MedChemExpress

Product Code. 16478257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478257 5 mg 5mg
16416116 100 mg 100mg
16415558 10 mg 10mg
16401727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478257 Supplier Medchem Express Supplier No. HY13209
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ambrisentan is a selective ET type A receptor (ETAR) antagonist.

Ambrisentan is a selective ET type A receptor (ETAR) antagonist.

Specifications

Chemical Name or Material Ambrisentan
Physical Form Solid
CAS 177036-94-1
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H22N2O4
For Use With (Application) COVID-19-immunoregulation
Synonym BSF 208075 LU 208075
Solubility Information DMSO : 100 mg/mL (264.26 mM; Need ultrasonic) ∣Ethanol : 7.14 mg/mL (18.87 mM; Need ultrasonic)
SMILES O=C([C@H](C(OC)(C1=CC=CC=C1)C2=CC=CC=C2)OC3=NC(C)=CC(C)=N3)O
Molecular Weight (g/mol) 378.42
Formula Weight 378.42
Percent Purity 99.58%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.