missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMDHD1 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 92%; Rabbit: 79%; Rat: 86%.
AMDHD1 is a protein coding gene.
Specifications
Specifications
| Antigen | AMDHD1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | AMDHD1 |
| Gene Accession No. | Q96NU7 |
| Gene Alias | 1300019J08Rik; Amdhd1; amidohydrolase domain containing 1; amidohydrolase domain-containing protein 1; HMFT1272; Probable imidazolonepropionase; RGD1308879 |
| Gene Symbols | AMDHD1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?