Learn More
Skytec™ AMF3CV P3 Disposable Respirator
Product Code. 30242979 Shop All Skytec Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
30242979 10/Pk. Pack of 10
1 options
Product Code. 30242979 Supplier Skytec™ Supplier No. MMFS/ALP/AMF3CV
  • ONLINE EXCLUSIVE PRICE
    114.12 €
    76.00 € / Pack of 10
    Save 38.12 €
    33% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Alpha Flow SERIES has been designed to improve comfort and performance whilst maintaining a secure fit. color coding the range makes identification simple and easy when varying levels of protection may be required.
  • Large surface area ensures very low breathing resistance
  • New large exhalation valve which reduces heat build-up and improves wearer comfort
  • color coded Alphamesh shell for ease of identification
  • Adjustable headstrap allows for a secure and comfortable fit
  • Metal-free - suitable for food industry applications
  • Preformed nosebridge eliminates the need for regular adjustment
  • Compatible with a wide range of safety eyewear
  • Soft flexible mesh maintains its shape and will not collapse during use
  • Conforms to EN 149: 2001 + A1: 2009 FFP3 NR
Specifications
Assigned Protection Factor (APF) 20
Certifications/Compliance CE , EN 149:2000 + A1:2009 FFP3 NR
Color Orange
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Filtration Layer Material Melt Blown Polypropylene
Material Headstrap Textile Elastic, Woven Elastic
Model Alphaflow
Quantity 10/Pk.
Sterility Non-sterile