missing translation for 'onlineSavingsMsg'
Learn More

Amifloxacin, MedChemExpress

Product Code. 16473239
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473239 1 mg 1mg
16422458 5 mg 5mg
16410259 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473239 Supplier Medchem Express Supplier No. HYU00221
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Amifloxacin (Win49375) is a synthetic antibacterial agent of the quinolone class.

Amifloxacin (Win49375) is a synthetic antibacterial agent of the quinolone class.

Specifications

Chemical Name or Material Amifloxacin
Physical Form Solid
CAS 86393-37-5
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C16H19FN4O3
For Use With (Application) COVID-19-immunoregulation
Synonym Win49375
Solubility Information H2O : 50 mg/mL (149.54 mM; ultrasonic and adjust pH to 12 with NaOH)
SMILES O=C(C1=CN(NC)C2=C(C=C(F)C(N3CCN(C)CC3)=C2)C1=O)O
Molecular Weight (g/mol) 334.35
Formula Weight 334.35
Percent Purity 99.23%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.