missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine hydrochloride, 98%

Product Code. 10354362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10354362 25 g Glass bottle 25g
10140812 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354362 Supplier Thermo Scientific Supplier No. 368910250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1937-19-5 | CH6N4·HCl | 110.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1937-19-5
Molecular Formula CH6N4·HCl
Molecular Weight (g/mol) 110.55
MDL Number MFCD00039074
InChI Key UBDZFAGVPPMTIT-UHFFFAOYSA-N
Synonym aminoguanidine hydrochloride, pimagedine hcl, guanylhydrazine hydrochloride, hydrazinecarboximidamide hydrochloride, aminoguanidinehydrochloride, aminoguanidine hcl, pimagedine hydrochloride, aminoguanidine monohydrochloride, hydrazinecarboximidamide, hydrochloride
PubChem CID 2734687
IUPAC Name 2-aminoguanidine;hydrochloride
SMILES C(=NN)(N)N.Cl

Specifications

Melting Point >160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NNHC(=NH)NH2·HCl
Quantity 25 g
Beilstein 03, IV, 236
Merck Index 15, 438
Solubility Information Solubility in water: soluble.
Formula Weight 110.55
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Aminoguanidine hydrochloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aminoguanidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.