accessibility menu, dialog, popup

UserName

CAS RN 1937-19-5

IUPAC Name: N''-azaniumylguanidine chloride
Synonyms: pimagedine
Molecular Weight (g/mol): 110.55
Molecular Formula: CH7ClN4
InChi Key: UBDZFAGVPPMTIT-UHFFFAOYSA-N
SMILES: [Cl-].NC(N)=N[NH3+]
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)